Assay Details
| Gene Symbol: pxna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201588.1 | NP_963882.1 | 6 - 7 | 1172 | 79 | |
| GenBank mRNA | AY515510.1 | - | 6 - 7 | 1172 | 79 | |
| BC054700.1 | - | 6 - 7 | 960 | 79 | ||
More Information
Set Membership: |
|

pxna
Genomic Map