Assay Details
| Gene Symbol: nr2f6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205557.1 | NP_991120.1 | 4 - 5 | 1317 | 68 | |
| XM_005171224.4 | XP_005171281.1 | 4 - 5 | 1561 | 68 | ||
| XM_005171225.4 | XP_005171282.1 | 4 - 5 | 1396 | 68 | ||
| XM_021478933.1 | XP_021334608.1 | 4 - 5 | 1455 | 68 | ||
| GenBank mRNA | BC065330.1 | - | 4 - 5 | 1317 | 68 | |
More Information
Set Membership: |
|

nr2f6a
Genomic Map