Assay Details
| Gene Symbol: arl8ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205566.1 | NP_991129.1 | 6 - 7 | 568 | 74 | |
| GenBank mRNA | BC065942.1 | - | 6 - 7 | 568 | 74 | |
More Information
Set Membership: |
|

arl8ba
Genomic Map