Assay Details
| Gene Symbol: dnaaf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205688.1 | NP_991251.1 | 1 - 2 | 143 | 96 | |
| GenBank mRNA | AY428510.1 | - | 1 - 2 | 239 | 96 | |
| BC065881.1 | - | 1 - 2 | 143 | 96 | ||
| BC165842.1 | - | 1 - 2 | 159 | 96 | ||
More Information
Set Membership: |
|

dnaaf4
Genomic Map