Assay Details
| Gene Symbol: dnaaf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205688.1 | NP_991251.1 | 4 - 5 | 659 | 72 | |
| GenBank mRNA | AY428510.1 | - | 4 - 5 | 755 | 72 | |
| BC065881.1 | - | 4 - 5 | 659 | 72 | ||
| BC165842.1 | - | 4 - 5 | 675 | 72 | ||
More Information
Set Membership: |
|

dnaaf4
Genomic Map