Assay Details
| Gene Symbol: irf1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205747.1 | NP_991310.1 | 7 - 8 | 748 | 116 | |
| GenBank mRNA | AY398364.1 | - | 7 - 8 | 748 | 116 | |
| BC085555.1 | - | 7 - 8 | 740 | 116 | ||
| BC165272.1 | - | 6 - 7 | 664 | 116 | ||
More Information
Set Membership: |
|

irf1b
Genomic Map