Assay Details
| Gene Symbol: vmp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207060.1 | NP_996943.1 | 10 - 11 | 1098 | 75 | |
| XM_005157413.4 | XP_005157470.1 | 11 - 12 | 1215 | 75 | ||
| GenBank mRNA | BC046904.1 | - | 11 - 12 | 1145 | 75 | |
| BC055534.1 | - | 11 - 12 | 1129 | 75 | ||
| BC066412.1 | - | 10 - 11 | 1098 | 75 | ||
More Information
Set Membership: |
|

vmp1
Genomic Map