Assay Details
| Gene Symbol: hnrnpaba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212587.3 | NP_997752.2 | 6 - 7 | 833 | 85 | |
| XM_005169577.2 | XP_005169634.1 | 6 - 7 | 960 | 85 | ||
| GenBank mRNA | BC048898.1 | - | 6 - 7 | 859 | 85 | |
| BC066454.1 | - | 6 - 7 | 833 | 85 | ||
More Information
Set Membership: |
|

hnrnpaba
Genomic Map