Assay Details
| Gene Symbol: hnrnpaba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212587.3 | NP_997752.2 | 8 - 9 | 1095 | 139 | |
| GenBank mRNA | BC048898.1 | - | 8 - 9 | 1121 | 139 | |
| BC066454.1 | - | 8 - 9 | 1095 | 139 | ||
More Information
Set Membership: |
|

hnrnpaba
Genomic Map