Assay Details
| Gene Symbol: tax1bp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212664.1 | NP_997829.1 | 12 - 13 | 1706 | 97 | |
| XM_005157956.3 | XP_005158013.1 | 12 - 13 | 1723 | 97 | ||
| XM_005157957.3 | XP_005158014.1 | 12 - 13 | 1792 | 97 | ||
| XM_021466620.1 | XP_021322295.1 | 13 - 14 | 1808 | 97 | ||
| XM_021466621.1 | XP_021322296.1 | 12 - 13 | 1795 | 97 | ||
| GenBank mRNA | BC060918.1 | - | 12 - 13 | 1723 | 97 | |
| BC065315.1 | - | 12 - 13 | 1706 | 97 | ||
More Information
Set Membership: |
|

tax1bp1b
Genomic Map