Assay Details
| Gene Symbol: tax1bp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212664.1 | NP_997829.1 | 14 - 15 | 1914 | 131 | |
| XM_005157957.3 | XP_005158014.1 | 14 - 15 | 2000 | 131 | ||
| XM_021466620.1 | XP_021322295.1 | 15 - 16 | 2016 | 131 | ||
| GenBank mRNA | BC065315.1 | - | 14 - 15 | 1914 | 131 | |
More Information
Set Membership: |
|

tax1bp1b
Genomic Map