Assay Details
| Gene Symbol: tax1bp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212664.1 | NP_997829.1 | 15 - 16 | 2087 | 119 | |
| XM_005157956.3 | XP_005158013.1 | 15 - 16 | 2101 | 119 | ||
| XM_005157957.3 | XP_005158014.1 | 15 - 16 | 2173 | 119 | ||
| XM_021466620.1 | XP_021322295.1 | 16 - 17 | 2189 | 119 | ||
| XM_021466621.1 | XP_021322296.1 | 14 - 15 | 2089 | 119 | ||
| GenBank mRNA | BC060918.1 | - | 15 - 16 | 2101 | 119 | |
| BC065315.1 | - | 15 - 16 | 2087 | 119 | ||
More Information
Set Membership: |
|

tax1bp1b
Genomic Map