Assay Details
| Gene Symbol: dnaja2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212665.1 | NP_997830.1 | 2 - 3 | 245 | 120 | |
| GenBank mRNA | BC045437.1 | - | 2 - 3 | 245 | 120 | |
| BC165837.1 | - | 2 - 3 | 165 | 120 | ||
More Information
Set Membership: |
|

dnaja2b
Genomic Map