Assay Details
| Gene Symbol: dnaja2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212665.1 | NP_997830.1 | 7 - 8 | 1040 | 85 | |
| GenBank mRNA | BC045437.1 | - | 7 - 8 | 1040 | 85 | |
| BC165837.1 | - | 7 - 8 | 960 | 85 | ||
More Information
Set Membership: |
|

dnaja2b
Genomic Map