Assay Details
| Gene Symbol: mknk2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212723.1 | NP_997888.1 | 13 - 14 | 1371 | 75 | |
| GenBank mRNA | BC045391.1 | - | 13 - 14 | 1371 | 75 | |
| BC165349.1 | - | 12 - 13 | 1189 | 75 | ||
More Information
Set Membership: |
|

mknk2a
Genomic Map