Assay Details
| Gene Symbol: mknk2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212723.1 | NP_997888.1 | 5 - 6 | 548 | 135 | |
| XM_021479017.1 | XP_021334692.1 | 5 - 6 | 565 | 135 | ||
| GenBank mRNA | BC045391.1 | - | 5 - 6 | 548 | 135 | |
| BC165349.1 | - | 4 - 5 | 366 | 135 | ||
More Information
Set Membership: |
|

mknk2a
Genomic Map