Assay Details
| Gene Symbol: mknk2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212723.1 | NP_997888.1 | 8 - 9 | 817 | 89 | |
| GenBank mRNA | BC045391.1 | - | 8 - 9 | 817 | 89 | |
| BC165349.1 | - | 7 - 8 | 635 | 89 | ||
More Information
Set Membership: |
|

mknk2a
Genomic Map