Assay Details
| Gene Symbol: cpn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212770.1 | NP_997935.1 | 5 - 6 | 881 | 64 | |
| GenBank mRNA | BC066689.1 | - | 5 - 6 | 881 | 64 | |
| BC154779.1 | - | 11 - 12 | 1283 | 64 | ||
More Information
Set Membership: |
|

cpn1
Genomic Map