Assay Details
| Gene Symbol: arglu1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213216.2 | NP_998381.2 | 2 - 3 | 707 | 148 | |
| GenBank mRNA | BC044383.1 | - | 2 - 3 | 707 | 148 | |
| BC062839.1 | - | 2 - 3 | 695 | 148 | ||
More Information
Set Membership: |
|

arglu1a
Genomic Map