Assay Details
| Gene Symbol: col9a1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213264.2 | NP_998429.2 | 16 - 17 | 893 | 91 | |
| GenBank mRNA | BC068419.1 | - | 16 - 17 | 861 | 91 | |
More Information
Set Membership: |
|

col9a1b
Genomic Map