Assay Details
| Gene Symbol: col9a1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213264.2 | NP_998429.2 | 18 - 19 | 1001 | 90 | |
| GenBank mRNA | BC068419.1 | - | 18 - 19 | 969 | 90 | |
More Information
Set Membership: |
|

col9a1b
Genomic Map