Assay Details
| Gene Symbol: aldh5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110468.1 | NP_001103938.2 | 8 | 1447 | 139 | |
| GenBank mRNA | BC134910.1 | - | 8 | 1447 | 139 | |
More Information
Set Membership: |
|

aldh5a1
Genomic Map