Assay Details
| Gene Symbol: tph1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001843.2 | NP_001001843.2 | 11 | 1848 | 136 | |
| GenBank mRNA | BC128802.1 | - | 11 | 1848 | 136 | |
| BC154120.1 | - | 11 | 1876 | 136 | ||
More Information
Set Membership: |
|

tph1b
Genomic Map