Assay Details
| Gene Symbol: pla2g12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082799.1 | NP_001076268.1 | 4 | 1448 | 145 | |
| GenBank mRNA | BC133875.1 | - | 4 | 1448 | 145 | |
More Information
Set Membership: |
|

pla2g12a
Genomic Map