Assay Details
| Gene Symbol: cmtm8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080645.1 | NP_001074114.1 | 4 | 1450 | 129 | |
| GenBank mRNA | BC129357.1 | - | 4 | 1450 | 129 | |
More Information
Set Membership: |
|

cmtm8b
Genomic Map