Assay Details
Gene Symbol: lhx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131218.1 | NP_571293.1 | 1 | 344 | 129 | |
GenBank mRNA | BC098523.1 | - | 1 | 373 | 129 | |
L42547.1 | - | 1 | 344 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
