Assay Details
| Gene Symbol: yod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017716.2 | NP_001017716.1 | 3 | 850 | 110 | |
| GenBank mRNA | BC093247.1 | - | 3 | 853 | 110 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

yod1| Gene Symbol: yod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017716.2 | NP_001017716.1 | 3 | 850 | 110 | |
| GenBank mRNA | BC093247.1 | - | 3 | 853 | 110 | |
Set Membership: |
|