Assay Details
| Gene Symbol: nrxn3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080009.1 | NP_001073478.1 | 22 | 4645 | 95 | |
| XM_021467191.1 | XP_021322866.1 | 23 | 4652 | 95 | ||
| XM_021467192.1 | XP_021322867.1 | 22 | 4628 | 95 | ||
| XM_021467193.1 | XP_021322868.1 | 22 | 4625 | 95 | ||
| XM_021467194.1 | XP_021322869.1 | 21 | 4610 | 95 | ||
| XM_021467195.1 | XP_021322870.1 | 20 | 4586 | 95 | ||
| XM_021467196.1 | XP_021322871.1 | 20 | 4583 | 95 | ||
| XM_021467197.1 | XP_021322872.1 | 19 | 4559 | 95 | ||
| XM_021467198.1 | XP_021322873.1 | 21 | 4610 | 95 | ||
| XM_021467199.1 | XP_021322874.1 | 21 | 4610 | 95 | ||
| XM_021467201.1 | XP_021322876.1 | 20 | 4583 | 95 | ||
| XM_021467202.1 | XP_021322877.1 | 20 | 4586 | 95 | ||
| XM_021467203.1 | XP_021322878.1 | 5 | 2281 | 95 | ||
| GenBank mRNA | DQ641432.1 | - | 22 | 4645 | 95 | |
| DQ641433.1 | - | 22 | 4645 | 95 | ||
| DQ641434.1 | - | 5 | 1494 | 95 | ||
More Information
Set Membership: |
|

nrxn3a
Genomic Map