Assay Details
| Gene Symbol: pi4k2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213358.1 | NP_998523.1 | 1749 | 107 | ||
| GenBank mRNA | BC058342.1 | - | 1749 | 107 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pi4k2a| Gene Symbol: pi4k2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213358.1 | NP_998523.1 | 1749 | 107 | ||
| GenBank mRNA | BC058342.1 | - | 1749 | 107 | ||
Set Membership: |
|