Assay Details
| Gene Symbol: gps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077759.1 | NP_001071227.1 | 14 | 1750 | 98 | |
| XM_021471590.1 | XP_021327265.1 | 14 | 1878 | 98 | ||
| GenBank mRNA | BC124805.1 | - | 14 | 1750 | 98 | |
More Information
Set Membership: |
|

gps1
Genomic Map