Assay Details
| Gene Symbol: rd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080631.1 | NP_001074100.1 | 3 | 1339 | 140 | |
| GenBank mRNA | BC129455.1 | - | 3 | 1339 | 140 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rd3| Gene Symbol: rd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080631.1 | NP_001074100.1 | 3 | 1339 | 140 | |
| GenBank mRNA | BC129455.1 | - | 3 | 1339 | 140 | |
Set Membership: |
|