Assay Details
| Gene Symbol: ar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083123.1 | NP_001076592.1 | 1 | 1752 | 154 | |
| GenBank mRNA | EF153102.1 | - | 1 | 154 | ||
| EF427915.1 | - | 1 | 1752 | 154 | ||
| EF440290.1 | - | 1 | 1745 | 154 | ||
| EU708622.1 | - | 1 | 1591 | 154 | ||
More Information
Set Membership: |
|

ar
Genomic Map