Assay Details
| Gene Symbol: c1r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030150.1 | NP_001025321.1 | 5 | 953 | 135 | |
| GenBank mRNA | BC096887.1 | - | 5 | 953 | 135 | |
| BC134118.1 | - | 10 | 2249 | 135 | ||
More Information
Set Membership: |
|

c1r
Genomic Map