Assay Details
| Gene Symbol: galnt7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020641.1 | NP_001018477.1 | 6 | 1148 | 125 | |
| GenBank mRNA | BC095642.1 | - | 6 | 1148 | 125 | |
More Information
Set Membership: |
|

galnt7
Genomic Map