Assay Details
| Gene Symbol: nrxn3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080047.1 | NP_001073516.1 | 18 | 4948 | 107 | |
| GenBank mRNA | DQ641435.1 | - | 18 | 4948 | 107 | |
| DQ641436.1 | - | 18 | 4948 | 107 | ||
| DQ641437.1 | - | 5 | 1915 | 107 | ||
More Information
Set Membership: |
|

nrxn3b
Genomic Map