Assay Details
| Gene Symbol: aurkaip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105695.1 | NP_001099165.1 | 5 | 749 | 105 | |
| GenBank mRNA | BC151937.1 | - | 5 | 749 | 105 | |
More Information
Set Membership: |
|

aurkaip1
Genomic Map