Assay Details
| Gene Symbol: zgc:174855 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103122.1 | NP_001096592.1 | 4 | 348 | 113 | |
| GenBank mRNA | BC152282.1 | - | 4 | 348 | 113 | |
| CT658773.2 | - | 3 | 236 | 113 | ||
| Gene Symbol: zgc:174154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001327979.1 | NP_001314908.1 | 4 | 348 | 113 | |
| GenBank mRNA | BC153505.1 | - | 4 | 348 | 113 | |
More Information
Set Membership: |
|

zgc:174855
Genomic Map