Assay Details
| Gene Symbol: polr3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200232.1 | NP_956526.1 | 2 | 148 | 66 | |
| GenBank mRNA | BC046092.1 | - | 2 | 148 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

polr3c| Gene Symbol: polr3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200232.1 | NP_956526.1 | 2 | 148 | 66 | |
| GenBank mRNA | BC046092.1 | - | 2 | 148 | 66 | |
Set Membership: |
|