Assay Details
| Gene Symbol: rps6ka3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083026.2 | NP_001076495.2 | 21 | 2842 | 154 | |
| XM_009297372.3 | XP_009295647.1 | 22 | 3037 | 154 | ||
| XM_009297376.3 | XP_009295651.1 | 21 | 2866 | 154 | ||
| XM_021470093.1 | XP_021325768.1 | 21 | 2711 | 154 | ||
| GenBank mRNA | BC133092.1 | - | 21 | 2843 | 154 | |
More Information
Set Membership: |
|

rps6ka3b
Genomic Map