Assay Details
| Gene Symbol: tp53bp2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214814.1 | NP_999979.1 | 12 | 1750 | 151 | |
| XM_005156475.4 | XP_005156532.1 | 12 | 1848 | 151 | ||
| XM_005156476.4 | XP_005156533.1 | 12 | 1834 | 151 | ||
| GenBank mRNA | BC070005.1 | - | 12 | 1750 | 151 | |
More Information
Set Membership: |
|

tp53bp2a
Genomic Map