Assay Details
| Gene Symbol: foxi3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198917.2 | NP_944599.2 | 2 | 1447 | 131 | |
| GenBank mRNA | BC133872.1 | - | 2 | 1447 | 131 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

foxi3a| Gene Symbol: foxi3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198917.2 | NP_944599.2 | 2 | 1447 | 131 | |
| GenBank mRNA | BC133872.1 | - | 2 | 1447 | 131 | |
Set Membership: |
|