Assay Details
| Gene Symbol: tsen15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100090.1 | NP_001093560.1 | 4 | 850 | 145 | |
| GenBank mRNA | BC161671.1 | - | 4 | 761 | 145 | |
More Information
Set Membership: |
|

tsen15
Genomic Map