Assay Details
| Gene Symbol: ywhag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213022.2 | NP_998187.1 | 2 | 324 | 119 | |
| GenBank mRNA | BC059494.1 | - | 2 | 247 | 119 | |
| EF095156.1 | - | 2 | 173 | 119 | ||
| EH437629.1 | - | 2 | 411 | 119 | ||
More Information
Set Membership: |
|

ywhag1
Genomic Map