Assay Details
| Gene Symbol: ednraa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099445.2 | NP_001092915.2 | 2 | 421 | 135 | |
| XM_021476949.1 | XP_021332624.1 | 3 | 490 | 135 | ||
| XM_021476969.1 | XP_021332644.1 | 2 | 307 | 135 | ||
| GenBank mRNA | BC167440.1 | - | 2 | 359 | 135 | |
| EF581835.1 | - | 1 | 154 | 135 | ||
| KM655667.1 | - | 1 | 115 | 135 | ||
More Information
Set Membership: |
|

ednraa
Genomic Map