Assay Details
| Gene Symbol: pkmyt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098196.1 | NP_001091666.1 | 8 | 1747 | 140 | |
| XM_005156122.4 | XP_005156179.1 | 8 | 1690 | 140 | ||
| GenBank mRNA | BC107967.1 | - | 8 | 1723 | 140 | |
| BC139690.1 | - | 8 | 1747 | 140 | ||
| BC160663.1 | - | 8 | 1744 | 140 | ||
More Information
Set Membership: |
|

pkmyt1
Genomic Map