Assay Details
| Gene Symbol: hnrnpub | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033595.2 | NP_001028767.2 | 13 | 4141 | 99 | |
| GenBank mRNA | AY570729.1 | - | 13 | 3951 | 99 | |
| CT598091.2 | - | 1 | 230 | 99 | ||
More Information
Set Membership: |
|

hnrnpub
Genomic Map