Assay Details
| Gene Symbol: prr15la | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003478.1 | NP_001003478.1 | 2 | 853 | 113 | |
| XM_021470304.1 | XP_021325979.1 | 3 | 1735 | 113 | ||
| GenBank mRNA | BC078363.1 | - | 2 | 853 | 113 | |
More Information
Set Membership: |
|

prr15la
Genomic Map