Assay Details
| Gene Symbol: galt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC129458.1 | - | 7 | 748 | 151 | |
| EH462716.1 | - | 1 | 498 | 151 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

galt| Gene Symbol: galt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC129458.1 | - | 7 | 748 | 151 | |
| EH462716.1 | - | 1 | 498 | 151 | ||
Set Membership: |
|