Assay Details
| Gene Symbol: prnpb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013297.2 | NP_001013315.2 | 2 | 1648 | 122 | |
| GenBank mRNA | AJ850286.2 | - | 2 | 1647 | 122 | |
| AY438683.1 | - | 2 | 1648 | 122 | ||
| JQ994489.1 | - | 1 | 1574 | 122 | ||
| KM655690.1 | - | 1 | 1574 | 122 | ||
More Information
Set Membership: |
|

prnpb
Genomic Map