Assay Details
| Gene Symbol: crb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044943.1 | NP_001038408.1 | 9 | 3247 | 146 | |
| GenBank mRNA | BC163555.1 | - | 9 | 3216 | 146 | |
| DQ314735.1 | - | 9 | 3247 | 146 | ||
More Information
Set Membership: |
|

crb1
Genomic Map